Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011874.2 | NP_036004.2 | 3 | 256 | ||
GenBank mRNA | AK050677.1 | - | 3 | 234 | ||
AK077507.1 | - | 3 | 219 | |||
AK150989.1 | - | 3 | 211 | |||
AK153991.1 | - | 3 | 239 | |||
AK160644.1 | - | 3 | 218 | |||
AK160718.1 | - | 3 | 210 | |||
AK167041.1 | - | 3 | 201 | |||
AK167528.1 | - | 3 | 211 | |||
AK169224.1 | - | 3 | 219 | |||
AK169244.1 | - | 3 | 719 | |||
BC012708.1 | - | 3 | 217 | 4211677 | ||
BC092265.1 | - | 3 | 200 | 3671250 | ||
DV646128.1 | - | 3 | 266 | |||
L76223.1 | - | 3 | 207 |