Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038231.2 | NP_001033320.1 | 3 | 482 | ||
NM_001286608.1 | NP_001273537.1 | 3 | 824 | |||
NM_011793.3 | NP_035923.1 | 3 | 690 | |||
GenBank mRNA | AB025349.1 | - | 3 | 422 | ||
AK003025.1 | - | 3 | 466 | |||
AK004212.1 | - | 3 | 449 | |||
AK010257.1 | - | 3 | 468 | |||
AK011079.1 | - | 3 | 471 | |||
AK011100.1 | - | 3 | 469 | |||
AK012588.1 | - | 3 | 452 | |||
AK088896.1 | - | 3 | 446 | |||
AK168039.1 | - | 3 | 471 | |||
AK185753.1 | - | 1 | 125 | |||
BC003709.1 | - | 3 | 459 | 3599828 | ||
CK780850.1 | - | 3 | 454 |