Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011801.1 | NP_035931.1 | 3 | 416 | ||
GenBank mRNA | AB010828.1 | - | 3 | 416 | ||
AK133989.1 | - | 3 | 434 | |||
AK160532.1 | - | 3 | 381 | |||
BC005589.1 | - | 3 | 397 | 3964401 | ||
Y08219.1 | - | 3 | 415 |