Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044389.2 | NP_001037854.1 | 9, 10 | 1491 | ||
NM_011802.3 | NP_035932.2 | 10, 11 | 1533 | |||
GenBank mRNA | AF134983.1 | - | 10, 11 | 1484 | ||
AJ276991.1 | - | 10, 11 | 1415 | |||
AK087848.1 | - | 10, 11 | 1487 | |||
BC061153.1 | - | 9, 10 | 1449 | 6593118 |