Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011814.2 | NP_035944.2 | 3 | 556 | ||
GenBank mRNA | AB025269.1 | - | 2 | 123 | ||
AK154243.1 | - | 3 | 555 | |||
AK172042.1 | - | 3 | 569 | |||
BC057007.1 | - | 3 | 451 | 6816943 | ||
BC062971.1 | - | 3 | 556 | 6415619 |