Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011823.4 | NP_035953.3 | 3 | 902 | ||
GenBank mRNA | AK021044.1 | - | 1 | 52 | ||
AK033675.1 | - | 3 | 884 | |||
AK039041.1 | - | 3 | 833 | |||
AK039528.1 | - | 3 | 881 | |||
AK140507.1 | - | 3 | 814 | |||
DQ103764.1 | - | 4 | 1046 | |||
DQ103765.1 | - | 5 | 1081 | |||
DQ103766.1 | - | 4 | 938 | |||
DQ103767.1 | - | 3 | 903 |
Set Membership: |
G-protein coupled receptor |