Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011826.4 | NP_035956.1 | 3, 4 | 636 | ||
GenBank mRNA | AF023482.1 | - | 3, 4 | 608 | ||
AF023483.1 | - | 2, 3 | 771 | |||
AK002256.1 | - | 3, 4 | 587 | |||
AK010633.1 | - | 3, 4 | 620 | |||
AK088746.1 | - | 3, 4 | 618 | |||
BC006688.1 | - | 3, 4 | 565 | 3582652 | ||
BC098225.1 | - | 3, 4 | 590 | 1494110 | ||
BC147481.1 | - | 3, 4 | 567 | 9056226 |