Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047537.1 | - | 5 | 548 | ||
AK089383.1 | - | 5 | 512 | |||
AK171139.1 | - | 5 | 523 | |||
AK199145.1 | - | 2 | 68 | |||
BC053416.1 | - | 4 | 372 | 6533681 | ||
CK619435.1 | - | 5 | 522 | |||
U00978.1 | - | 4 | 267 |