Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011834.2 | NP_035964.1 | 13 | 1742 | ||
GenBank mRNA | AF072376.1 | - | 13 | 1742 |