Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011840.2 | NP_035970.1 | 22 | 2151 | ||
GenBank mRNA | AB019374.1 | - | 22 | 2152 | ||
AK159261.1 | - | 22 | 2201 | |||
BC028260.1 | - | 21 | 2004 | 5067818 |