Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099067.1 | - | 6 | 1040 | ||
AJ223071.1 | - | 6 | 1130 | |||
AK078809.1 | - | 6 | 1152 | |||
AK132803.1 | - | 6 | 1068 | |||
AK144130.1 | - | 6 | 1014 | |||
AK147381.1 | - | 6 | 1118 | |||
AK149584.1 | - | 6 | 1068 | |||
BC057939.1 | - | 6 | 1042 | 5325558 | ||
CN526970.1 | - | 1 | 36 | |||
Y09234.1 | - | 6 | 1130 |