Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290707.1 | NP_001277636.1 | 12, 13 | 1378 | ||
NM_011866.2 | NP_035996.2 | 11, 12 | 1332 | |||
GenBank mRNA | AF110507.1 | - | 11, 12 | 1203 | ||
AK039249.1 | - | 12, 13 | 1378 | |||
AK147690.1 | - | 10, 11 | 1249 | |||
AK162221.1 | - | 11, 12 | 987 | |||
AK162804.1 | - | 9, 10 | 873 | |||
AK166310.1 | - | 3, 4 | 344 | |||
AY360383.1 | - | 11, 12 | 1332 | |||
BC113201.1 | - | 11, 12 | 1198 | 40048238 |