Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011867.3 | NP_035997.1 | 21 | 3028 | ||
GenBank mRNA | AF167411.1 | - | 21 | 3029 |