Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000742.3 | NP_000733.2 | 5 | 1018 | ||
NM_001282455.1 | NP_001269384.1 | 5 | 973 | |||
XM_005273397.2 | XP_005273454.1 | 5 | 942 | |||
XM_006716282.1 | XP_006716345.1 | 6 | 929 | |||
XM_011544388.1 | XP_011542690.1 | 6 | 1020 | |||
XM_011544389.1 | XP_011542691.1 | 5 | 1018 | |||
XM_017013003.1 | XP_016868492.1 | 5 | 950 | |||
XM_017013004.1 | XP_016868493.1 | 6 | 884 | |||
GenBank mRNA | AK296348.1 | - | 5 | 528 | ||
AK308465.1 | - | 1 | 249 | |||
AK314787.1 | - | 5 | 936 | |||
BC153866.1 | - | 4 | 444 | 40121686 | ||
CD013901.1 | - | 4 | 584 | |||
DA096824.1 | - | 4 | 410 | |||
U62431.1 | - | 5 | 963 | |||
Y16281.1 | - | 4 | 409 |