Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011886.2 | NP_036016.2 | 4, 5 | 506 | ||
GenBank mRNA | AF005036.1 | - | 4, 5 | 413 | ||
AF295403.1 | - | 4, 5 | 411 | |||
AK135335.1 | - | 4, 5 | 490 | |||
AK146204.1 | - | 4, 5 | 508 | |||
AK160549.1 | - | 4, 5 | 491 | |||
AK170115.1 | - | 4, 5 | 473 | |||
BC002021.1 | - | 4, 5 | 504 | 3489293 |