Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145858.1 | NP_001139330.1 | 2 | 317 | ||
NM_001145859.1 | NP_001139331.1 | 2 | 320 | |||
NM_011893.3 | NP_036023.3 | 2 | 217 | |||
GenBank mRNA | AK089455.1 | - | 2 | 148 | ||
AK143548.1 | - | 2 | 322 | |||
AK150265.1 | - | 2 | 319 | |||
AK155103.1 | - | 2 | 174 | |||
AK155898.1 | - | 2 | 217 | |||
AK155970.1 | - | 2 | 265 | |||
AK156912.1 | - | 2 | 262 | |||
BC010198.1 | - | 2 | 155 | 3967543 | ||
BC085497.1 | - | 2 | 208 | 5696296 | ||
L14543.1 | - | 2 | 151 |