Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011933.2 | NP_036063.1 | 1, 2 | 210 | ||
GenBank mRNA | AF155575.1 | - | 1, 2 | 210 | ||
AK137212.1 | - | 1, 2 | 141 | |||
AK149270.1 | - | 1, 2 | 335 | |||
AK162961.1 | - | 1, 2 | 346 | |||
BC021865.1 | - | 1, 2 | 182 | 5123899 |