Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011948.2 | NP_036078.2 | 2 | 226 | ||
GenBank mRNA | AK014185.1 | - | 2 | 345 | ||
AK035823.1 | - | 2 | 363 | |||
AK133605.1 | - | 2 | 363 | |||
BC058719.1 | - | 2 | 226 | 5705378 | ||
U85607.1 | - | 2 | 314 | |||
U85608.1 | - | 2 | 314 |