Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011956.3 | NP_036086.1 | 2, 3 | 199 | ||
GenBank mRNA | AF114169.1 | - | 2, 3 | 177 | ||
AK003816.1 | - | 2, 3 | 197 | |||
AK169896.1 | - | 2, 3 | 158 | |||
BC012635.1 | - | 2, 3 | 200 | 4038123 | ||
BI735707.1 | - | 2, 3 | 199 |