Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011960.2 | NP_036090.2 | 2 | 545 | ||
GenBank mRNA | AF079557.1 | - | 2 | 294 | ||
AK036656.1 | - | 2 | 568 | |||
AK080306.1 | - | 2 | 265 | |||
AK160742.1 | - | 1 | 19 | |||
AK164013.1 | - | 2 | 532 | |||
BC050892.1 | - | 2 | 544 | |||
BC053037.1 | - | 2 | 528 | |||
BC059827.1 | - | 2 | 554 |