Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289476.1 | NP_001276405.1 | 2 | 847 | ||
NM_011969.2 | NP_036099.1 | 2 | 208 | |||
GenBank mRNA | AF019662.1 | - | 2 | 146 | ||
AK088615.1 | - | 2 | 153 | |||
AK146765.1 | - | 2 | 165 | |||
AK165563.1 | - | 2 | 161 | |||
AK168234.1 | - | 2 | 179 | |||
BC008222.1 | - | 2 | 141 | 2581897 | ||
BC014752.1 | - | 2 | 795 | |||
BU841938.1 | - | 2 | 211 | |||
BY779773.1 | - | 2 | 196 | |||
DV049235.1 | - | 2 | 177 |