Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 2, 3 | 213 | ||
AF034568.1 | - | 2, 3 | 267 | |||
AK002922.1 | - | 2, 3 | 316 | |||
AK079488.1 | - | 2, 3 | 313 | |||
AK142016.1 | - | 2, 3 | 310 | |||
AK150497.1 | - | 2, 3 | 320 | |||
AK151463.1 | - | 2, 3 | 334 | |||
AK160007.1 | - | 2, 3 | 322 | |||
AK160706.1 | - | 2, 3 | 300 | |||
AK169305.1 | - | 2, 3 | 286 | |||
BC043657.1 | - | 2, 3 | 285 | 5343539 |