Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011902.2 | NP_036032.2 | 4 | 526 | ||
GenBank mRNA | AB027138.1 | - | 4 | 526 | ||
AK006205.1 | - | 3 | 273 | |||
BC008140.1 | - | 4 | 542 | 3582971 | ||
BQ712964.1 | - | 4 | 540 |