Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 9 | 1228 | ||
NM_001294329.1 | NP_001281258.1 | 7 | 1088 | |||
NM_001294330.1 | NP_001281259.1 | 7 | 1036 | |||
NM_001294331.1 | NP_001281260.1 | 9 | 1241 | |||
NM_001294334.1 | NP_001281263.1 | 9 | 1228 | |||
GenBank mRNA | AF045252.1 | - | 9 | 1247 | ||
AF045253.1 | - | 8 | 675 | |||
AF045254.1 | - | 4 | 389 | |||
AK014829.1 | - | 9 | 866 | |||
AK089925.1 | - | 7 | 616 | |||
AK165667.1 | - | 9 | 1231 | |||
AX775743.1 | - | 3 | 253 | |||
BC066198.1 | - | 7 | 581 | |||
CD554252.1 | - | 3 | 352 |