Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011921.2 | NP_036051.1 | 7, 8 | 799 | ||
GenBank mRNA | BC032880.1 | - | 7, 8 | 776 | ||
BC046315.1 | - | 7, 8 | 773 | 6314273 | ||
BC139412.1 | - | 7, 8 | 776 | 8862434 | ||
BC139413.1 | - | 7, 8 | 776 | 8862435 | ||
U96401.1 | - | 7, 8 | 799 |