Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136089.2 | NP_001129561.1 | 4, 5 | 482 | ||
NM_011922.3 | NP_036052.2 | 4, 5 | 482 | |||
GenBank mRNA | AJ238978.1 | - | 4, 5 | 382 | ||
AK008808.1 | - | 4, 5 | 406 | |||
AK018597.1 | - | 4, 5 | 439 | |||
AK020288.1 | - | 4, 5 | 407 | |||
AK131880.1 | - | 4, 5 | 402 | |||
AK168715.1 | - | 4, 5 | 386 | |||
BC125319.1 | - | 4, 5 | 354 | 40129834 | ||
BC132207.1 | - | 4, 5 | 354 | 40130484 |