Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011932.2 | NP_036062.1 | 4 | 530 | ||
GenBank mRNA | AF163255.1 | - | 4 | 484 | ||
AF186023.1 | - | 4 | 505 | |||
AK076886.1 | - | 2 | 83 | |||
AK145745.1 | - | 4 | 496 | |||
AK146180.1 | - | 4 | 499 | |||
AK169584.1 | - | 3 | 395 | |||
BC014759.1 | - | 4 | 526 | 4486834 |