Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000831.3 | NP_000822.2 | 2 | 311 | ||
GenBank mRNA | AJ249210.1 | - | 2 | 197 | ||
AJ299451.1 | - | 2 | 176 | |||
BI544689.1 | - | 315 | ||||
DR003075.1 | - | 2 | 495 | |||
U16127.1 | - | 2 | 194 |