Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011956.3 | NP_036086.1 | 3, 4 | 383 | ||
GenBank mRNA | AF114169.1 | - | 3, 4 | 361 | ||
AK003816.1 | - | 3, 4 | 381 | |||
AK169896.1 | - | 3, 4 | 342 | |||
AK210395.1 | - | 1, 2 | 167 | |||
AK213247.1 | - | 1, 2 | 166 | |||
BC012635.1 | - | 3, 4 | 384 | 4038123 | ||
BI735707.1 | - | 3, 4 | 383 |