Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011978.2 | NP_036108.2 | 2 | 661 | ||
GenBank mRNA | AF033031.1 | - | 2 | 542 | ||
AF072757.1 | - | 2 | 530 | |||
AJ223958.1 | - | 2 | 579 | |||
AK144069.1 | - | 2 | 663 | |||
AK144070.1 | - | 2 | 575 | |||
AK165446.1 | - | 2 | 662 | |||
BC013442.1 | - | 2 | 644 | 4236616 | ||
BC022170.1 | - | 2 | 640 | 4159081 | ||
BC024735.1 | - | 2 | 630 | 5123650 |