Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011990.2 | NP_036120.1 | 1 | 471 | ||
GenBank mRNA | AB022345.1 | - | 1 | 438 | ||
AK037490.1 | - | 1 | 472 | |||
AK037742.1 | - | 1 | 446 | |||
AK076322.1 | - | 1 | 445 | |||
AK089202.1 | - | 1 | 444 | |||
AK089283.1 | - | 1 | 463 | |||
AK162233.1 | - | 1 | 244 | |||
AK163168.1 | - | 1 | 445 | |||
AK165599.1 | - | 1 | 447 | |||
AK170568.1 | - | 1 | 340 | |||
AK171192.1 | - | 1 | 469 | |||
AK171252.1 | - | 1 | 445 | |||
AK171327.1 | - | 1 | 447 | |||
AY766236.1 | - | 1 | 438 | |||
AY766237.1 | - | 438 | ||||
BB630675.1 | - | 1 | 472 | |||
BC141402.1 | - | 1 | 368 | 9056015 | ||
BC141408.1 | - | 1 | 368 | 9056021 |