Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011889.2 | NP_036019.2 | 7 | 834 | ||
GenBank mRNA | AF104411.1 | - | 7 | 857 | ||
AK030668.1 | - | 7 | 802 | |||
AK046722.1 | - | 7 | 927 | |||
AK141915.1 | - | 7 | 761 | |||
AK164861.1 | - | 7 | 931 | |||
AK169916.1 | - | 7 | 955 | |||
AK220520.1 | - | 5 | 633 | |||
BC055738.1 | - | 7 | 763 | 6413697 |