Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011894.2 | NP_036024.2 | 9 | 1414 | ||
GenBank mRNA | AB016835.1 | - | 9 | 1245 | ||
AK043375.1 | - | 1459 | ||||
AK161427.1 | - | 8 | 1396 | |||
AK165033.1 | - | 9 | 1450 | |||
AK219620.1 | - | 1 | 42 | |||
BC018237.1 | - | 9 | 1446 | 3482670 | ||
BC053741.1 | - | 9 | 1414 | 6336511 |