Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 13 | 1687 | ||
NM_001294329.1 | NP_001281258.1 | 11 | 1547 | |||
NM_001294330.1 | NP_001281259.1 | 11 | 1495 | |||
NM_001294331.1 | NP_001281260.1 | 13 | 1700 | |||
NM_001294334.1 | NP_001281263.1 | 13 | 1687 | |||
GenBank mRNA | AF045252.1 | - | 13 | 1706 | ||
AF045253.1 | - | 12 | 1134 | |||
AF045254.1 | - | 8 | 848 | |||
AK014829.1 | - | 13 | 1325 | |||
AK089925.1 | - | 11 | 1075 | |||
AK165667.1 | - | 13 | 1690 | |||
AX775743.1 | - | 7 | 712 | |||
BC066198.1 | - | 11 | 1040 | |||
U56909.1 | - | 4 | 385 |