Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011907.4 | NP_036037.1 | 2 | 968 | ||
GenBank mRNA | AF151108.1 | - | 2 | 955 |