Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011916.2 | NP_036046.2 | 41, 42 | 4992 | ||
GenBank mRNA | AK052194.1 | - | 8, 9 | 963 | ||
AK169840.1 | - | 9, 10 | 1091 | |||
BC054108.1 | - | 39, 40 | 5728 | |||
D88026.1 | - | 41, 42 | 4883 | |||
X91617.1 | - | 41, 42 | 4973 |