Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011937.2 | NP_036067.2 | 7 | 921 | ||
GenBank mRNA | AF088903.1 | - | 7 | 879 | ||
AF160355.1 | - | 7 | 867 | |||
AK077625.1 | - | 4 | 586 | |||
AK080610.1 | - | 7 | 870 | |||
AK166069.1 | - | 7 | 872 | |||
AK167088.1 | - | 4 | 595 | |||
AK172879.1 | - | 6 | 784 | |||
BC014800.1 | - | 7 | 852 | 4220980 |
Gene Symbol: Gm8615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB237974.1 | - | 1 | 100 |