Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011948.2 | NP_036078.2 | 21 | 4061 | ||
GenBank mRNA | AK122219.1 | - | 20 | 3744 | ||
BC058719.1 | - | 21 | 4061 | 5705378 | ||
U85607.1 | - | 21 | 4149 | |||
U85608.1 | - | 20 | 3993 |