Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011986.4 | NP_036116.3 | 3 | 1115 | ||
GenBank mRNA | AK122327.1 | - | 1 | 460 | ||
AK154889.1 | - | 3 | 1096 | |||
AK170502.1 | - | 4 | 830 | |||
AK216583.1 | - | 1 | 207 | |||
BC017126.1 | - | 3 | 1046 | 4485839 | ||
BQ745150.1 | - | 2 | 545 | |||
CX568560.1 | - | 1 | 501 |