Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011990.2 | NP_036120.1 | 12 | 2229 | ||
GenBank mRNA | AB022345.1 | - | 12 | 2196 | ||
AK076322.1 | - | 12 | 2204 | |||
AK162233.1 | - | 12 | 2004 | |||
AK165599.1 | - | 12 | 2205 | |||
AK171192.1 | - | 12 | 2227 | |||
AK171252.1 | - | 12 | 2203 | |||
AY766236.1 | - | 12 | 2196 | |||
BC141402.1 | - | 12 | 2126 | 9056015 | ||
BC141408.1 | - | 12 | 2126 | 9056021 |