Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122978.1 | NP_001116450.1 | 3 | 138 | ||
NM_011997.2 | NP_036127.2 | 3 | 385 | |||
GenBank mRNA | AF132726.1 | - | 3 | 345 | ||
AK142148.1 | - | 3 | 137 | |||
AK165109.1 | - | 3 | 139 | |||
BC138022.1 | - | 2 | 128 | 8861041 | ||
BC138041.1 | - | 2 | 128 | 8861060 |