Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169.2 | NP_001160.2 | 4 | 497 | ||
XM_011545822.2 | XP_011544124.1 | 4 | 540 | |||
GenBank mRNA | AB013456.1 | - | 4 | 499 | ||
AF067797.1 | - | 4 | 500 | |||
AK222964.1 | - | 4 | 462 | |||
AK222975.1 | - | 4 | 494 | |||
BC040630.1 | - | 4 | 465 | 5759105 |