Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 1, 2 | 462 | ||
NM_201245.3 | NP_957697.1 | 1, 2 | 462 | |||
GenBank mRNA | AK044512.1 | - | 1, 2 | 212 | ||
AK155630.1 | - | 1, 2 | 243 | |||
AK158296.1 | - | 1, 2 | 451 | |||
AK170849.1 | - | 1, 2 | 446 | |||
AK171089.1 | - | 1, 2 | 451 | |||
BC049803.1 | - | 1, 2 | 102 | |||
BC063067.1 | - | 1, 2 | 425 | 5721396 | ||
U73200.1 | - | 1, 2 | 322 |