Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecsit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253897.1 | NP_001240826.1 | 4 | 434 | ||
NM_001253898.1 | NP_001240827.1 | 3 | 256 | |||
NM_012029.2 | NP_036159.1 | 4 | 442 | |||
GenBank mRNA | AF182510.1 | - | 3 | 455 | ||
AK007564.1 | - | 4 | 416 | |||
AK012835.1 | - | 4 | 414 | |||
BC004583.1 | - | 3 | 243 | 3587327 | ||
BC016424.1 | - | 4 | 382 | 4924678 | ||
BY123162.1 | - | 3 | 236 | |||
CB589832.1 | - | 2 | 138 |