Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 5 | 672 | ||
NM_001318156.1 | NP_001305085.1 | 4 | 554 | |||
NM_001318157.1 | NP_001305086.1 | 5 | 990 | |||
NM_001318158.1 | NP_001305087.1 | 5 | 672 | |||
NR_134513.1 | 4 | 548 | ||||
NR_134514.1 | 4 | 548 | ||||
NR_134515.1 | 5 | 699 | ||||
XM_005251453.3 | XP_005251510.1 | 6 | 772 | |||
XM_006716765.3 | XP_006716828.1 | 4 | 433 | |||
XM_011517866.2 | XP_011516168.1 | 5 | 582 | |||
XM_011517867.2 | XP_011516169.1 | 3 | 212 | |||
XM_017014700.1 | XP_016870189.1 | 5 | 873 | |||
XM_017014701.1 | XP_016870190.1 | 4 | 543 | |||
XM_017014702.1 | XP_016870191.1 | 5 | 578 | |||
XM_017014703.1 | XP_016870192.1 | 4 | 331 | |||
XM_017014704.1 | XP_016870193.1 | 3 | 639 | |||
XM_017014705.1 | XP_016870194.1 | 3 | 309 | |||
XM_017014706.1 | XP_016870195.1 | 6 | 703 | |||
GenBank mRNA | AB006190.1 | - | 5 | 509 | ||
AK294028.1 | - | 5 | 483 | |||
AK297860.1 | - | 5 | 577 | |||
AK297885.1 | - | 5 | 806 | |||
AK300716.1 | - | 2 | 450 | |||
AK301949.1 | - | 4 | 511 | |||
AK310456.1 | - | 3 | 411 | |||
AK315879.1 | - | 2 | 441 | |||
BC062701.1 | - | 4 | 381 | 30324263 | ||
BC119672.1 | - | 4 | 380 | |||
BC119673.1 | - | 4 | 380 |