Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013475.4 | NP_038503.4 | 2 | 233 | ||
GenBank mRNA | AK010926.1 | - | 2 | 221 | ||
BC011037.1 | - | 2 | 249 | |||
BC019811.1 | - | 2 | 221 | 5125770 | ||
BC053338.1 | - | 2 | 209 | 5125963 | ||
BY143138.1 | - | 2 | 233 | |||
D10056.1 | - | 2 | 207 | |||
S70439.1 | - | 2 | 217 | |||
Y11356.1 | - | 2 | 237 |