Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048208.1 | NP_001041673.1 | 4, 5 | 821 | ||
NM_012005.3 | NP_036135.3 | 4, 5 | 821 | |||
GenBank mRNA | AB019028.1 | - | 4, 5 | 819 | ||
AK084044.1 | - | 4, 5 | 678 | |||
AK163306.1 | - | 5, 6 | 583 | |||
BC065072.1 | - | 4, 5 | 921 | 6830820 |