Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012006.2 | NP_036136.1 | 3 | 874 | ||
GenBank mRNA | AK053680.1 | - | 887 | |||
BC003911.1 | - | 3 | 871 | |||
BC022951.1 | - | 2 | 1924 | |||
BC028261.1 | - | 3 | 884 | 5100478 | ||
BC040749.1 | - | 2 | 1924 | |||
Y14004.1 | - | 3 | 874 |