Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012021.2 | NP_036151.1 | 2 | 273 | ||
GenBank mRNA | AB041573.1 | - | 2 | 606 | ||
AF110733.1 | - | 2 | 199 | |||
AF124994.1 | - | 2 | 362 | |||
AF197951.1 | - | 2 | 273 | |||
AK002383.1 | - | 2 | 239 | |||
AK003332.2 | - | 2 | 239 | |||
AK009858.1 | - | 2 | 708 | |||
AK136130.1 | - | 2 | 214 | |||
AK152653.1 | - | 2 | 239 | |||
AK209224.1 | - | 1 | 43 | |||
BC008174.1 | - | 2 | 658 | 3481713 |