Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK173325.1 | - | 4 | 330 | ||
BC022978.1 | - | 4 | 235 | |||
BC057883.1 | - | 4 | 275 | |||
BC075734.1 | - | 4 | 295 | |||
BC138030.1 | - | 4 | 231 | 8861049 | ||
U73199.1 | - | 4 | 307 |